LIF432341719 Life Chemicals Inc.

(5-Aminomethyl-pyridin-2-yl)-diethyl-amine

  • MDL number MFCD09044973
  • Cas Number : 926234-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart