LIF432341723 Life Chemicals Inc.

(2-methoxypyridin-4-yl)methanamine

  • MDL number MFCD09054679
  • Cas Number : 148900-69-0
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart