LIF432341726 Life Chemicals Inc.

[2-(2-methoxyethoxy)pyridin-4-yl]methanamine

  • MDL number MFCD09815188
  • Cas Number : 1016496-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart