LIF432341727 Life Chemicals Inc.

[(6-methoxypyridin-3-yl)methyl]amine

  • MDL number MFCD06212825
  • Cas Number : 262295-96-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart