LIF432341729 Life Chemicals Inc.

[6-(2-methoxyethoxy)pyridin-3-yl]methanamine

  • MDL number MFCD09806953
  • Cas Number : 1016724-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart