LIF432341732 Life Chemicals Inc.

6-ethoxynicotinic acid

  • MDL number MFCD07403514
  • Cas Number : 97455-65-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart