LIF432341734 Life Chemicals Inc.

3-(bromomethyl)-1,1-dimethylcyclobutane

  • MDL number MFCD17170399
  • Cas Number : 76207-22-2
  • Molecular Weight : 177.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart