LIF432341740 Life Chemicals Inc.

2-(1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD08701066
  • Cas Number : 956722-93-3
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart