LIF432341751 Life Chemicals Inc.

[2-(4-methylpiperazin-1-yl)ethyl]amine trihydrochloride

  • MDL number MFCD08437659
  • Cas Number : 90482-07-8
  • Molecular Weight : 252.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart