LIF432341752 Life Chemicals Inc.

2-(4-chlorophenyl)-2-phenylacetic acid

  • MDL number MFCD01631956
  • Cas Number : 21771-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart