LIF432341755 Life Chemicals Inc.

N-cyclohexyl-N-methylethane-1,2-diamine dihydrochloride

  • MDL number MFCD11808690
  • Cas Number : 1177346-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart