LIF432341756 Life Chemicals Inc.

(3-phenoxybenzyl)amine hydrochloride

  • MDL number MFCD07781045
  • Cas Number : 376637-85-3
  • Molecular Weight : 235.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart