LIF432341757 Life Chemicals Inc.

[1-(3-methoxyphenyl)propyl]methylamine hydrochloride

  • MDL number MFCD10083097
  • Cas Number : 1093405-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart