LIF432341760 Life Chemicals Inc.

[2-(4-methyl-1,3-thiazol-2-yl)ethyl]amine dihydrochloride

  • MDL number MFCD10083099
  • Cas Number : 1134723-32-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart