LIF432341762 Life Chemicals Inc.

N-methyl-2-pyrrolidin-1-ylethanamine dihydrochloride

  • MDL number MFCD20526814
  • Cas Number : 1051364-49-8
  • Molecular Weight : 201.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart