LIF432341766 Life Chemicals Inc.

[3-(1,3-thiazol-2-yl)benzyl]amined dihydrochloride

  • MDL number MFCD19691836
  • Cas Number : 1276631-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart