LIF432341767 Life Chemicals Inc.

[4-(1,3-thiazol-2-yl)benzyl]amine hydrochloride

  • MDL number MFCD14525839
  • Cas Number : 1187451-28-0
  • Molecular Weight : 226.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart