LIF432341770 Life Chemicals Inc.

[3-(6,7-dimethoxy-3,4-dihydroisoquinolin-2(1H)-yl)propyl]amine dihydrochloride

  • MDL number MFCD22587883
  • Cas Number : 93808-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart