LIF432341777 Life Chemicals Inc.

4-(2-Methyl-imidazol-1-ylmethyl)-benzoic acid hydrochloride

  • MDL number MFCD09743476
  • Cas Number : 1134483-46-7
  • Molecular Weight : 252.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart