LIF432341778 Life Chemicals Inc.

1-(cyclopropylcarbonyl)piperazine hydrochloride

  • MDL number MFCD03428576
  • Cas Number : 1021298-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart