LIF432341790 Life Chemicals Inc.

(1-methyl-1H-benzo[d]imidazol-2-yl)methanamine dihydrochloride

  • MDL number MFCD06687785
  • Cas Number : 53332-79-9
  • Molecular Weight : 234.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart