LIF432341793 Life Chemicals Inc.

4-[(2,6-dimethylmorpholin-4-yl)methyl]-1,3-thiazol-2-amine dihydrochloride

  • MDL number MFCD16661090
  • Cas Number : 1426291-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart