LIF432341797 Life Chemicals Inc.

N-(4-methyl-1,3-thiazol-2-yl)piperidine-4-carboxamide hydrochloride

  • MDL number MFCD16661119
  • Cas Number : 1351643-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart