LIF432341798 Life Chemicals Inc.

[4-(prop-2-yn-1-yloxy)phenyl]amine hydrochloride

  • MDL number MFCD18434594
  • Cas Number : 1217129-50-4
  • Molecular Weight : 183.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart