LIF432341801 Life Chemicals Inc.

3,5-dimethyl-1-(propan-2-yl)-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD12403822
  • Cas Number : 1183269-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart