LIF432341807 Life Chemicals Inc.

4-(2-amino-2-oxoethoxy)benzenesulfonyl chloride

  • MDL number MFCD08457512
  • Cas Number : 69986-21-6
  • Molecular Weight : 249.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart