LIF432341810 Life Chemicals Inc.

1-methyl-2-(propan-2-yl)-1H-imidazole-4-sulfonyl chloride

  • MDL number MFCD19663586
  • Cas Number : 1340300-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart