LIF432341815 Life Chemicals Inc.

5-(isocyanatomethyl)-2H-1,3-benzodioxole

  • MDL number MFCD08435909
  • Cas Number : 71217-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart