LIF432341817 Life Chemicals Inc.

3-methanesulfonamidopropanoic acid

  • MDL number MFCD06434734
  • Cas Number : 105611-92-5
  • Molecular Weight : 167.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart