LIF432341826 Life Chemicals Inc.

1-(5-Methoxy-2-methyl-1-benzofuran-3-yl)ethanamine hydrochloride

  • MDL number MFCD06409289
  • Cas Number : 1182284-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart