LIF432341831 Life Chemicals Inc.

3-(ethanesulfonyl)azetidine hydrochloride

  • MDL number MFCD26792957
  • Cas Number : 1820707-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart