LIF432341832 Life Chemicals Inc.

3-[(4-chlorophenyl)sulfonyl]azetidine hydrochloride

  • MDL number MFCD26792960
  • Cas Number : 1820665-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart