LIF432341833 Life Chemicals Inc.

3-((4-chlorophenyl)sulfonyl)pyrrolidine hydrochloride

  • MDL number MFCD28016977
  • Cas Number : 101768-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart