LIF432341835 Life Chemicals Inc.

1-methyl-5-(pyrrolidin-3-ylsulfanyl)-1H-1,2,3,4-tetrazole hydrochloride

  • MDL number MFCD26793085
  • Cas Number : 1820608-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart