LIF432341836 Life Chemicals Inc.

4-((4-fluorophenyl)thio)piperidine hydrochloride

  • MDL number MFCD04117760
  • Cas Number : 101798-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart