LIF432341844 Life Chemicals Inc.

4-(2-((ethylthio)methyl)pyrrolidin-1-yl)piperidine

  • MDL number MFCD28016979
  • Cas Number : 1706429-22-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart