LIF432341847 Life Chemicals Inc.

2-methylpyrazolo[1,5-a]pyrimidine

  • MDL number MFCD08272070
  • Cas Number : 78562-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart