LIF432341848 Life Chemicals Inc.

2-(pyrrolidin-3-yl)-1,2,3,4-tetrahydroisoquinoline dihydrochloride

  • MDL number MFCD13561602
  • Cas Number : 1220017-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart