LIF432341849 Life Chemicals Inc.

5-(piperidin-4-yl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine dihydrochloride

  • MDL number MFCD28016994
  • Cas Number : 1820650-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart