LIF432341870 Life Chemicals Inc.

thiazolo[3,2-b][1,2,4]triazol-6-ylmethanamine

  • MDL number MFCD22587928
  • Cas Number : 1219905-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart