LIF432341875 Life Chemicals Inc.

4-acetamido-2-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD04117318
  • Cas Number : 62374-67-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart