LIF432341885 Life Chemicals Inc.

N-methyl-1-(3-methyl-1,2,4-oxadiazol-5-yl)methanamine hydrochloride

  • MDL number MFCD27977456
  • Cas Number : 1573547-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart