LIF432341886 Life Chemicals Inc.

1-(3-isopropyl-1,2,4-oxadiazol-5-yl)-N-methylmethanamine hydrochloride

  • MDL number MFCD27977457
  • Cas Number : 1573547-75-7
  • Molecular Weight : 191.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart