LIF432341898 Life Chemicals Inc.

(2-pyrrolidin-1-ylpropyl)amine

  • MDL number MFCD06446933
  • Cas Number : 50998-07-7
  • Molecular Weight : 128.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart