LIF432341899 Life Chemicals Inc.

(3-azepan-1-ylpropyl)amine dihydrochloride

  • MDL number MFCD07371477
  • Cas Number : 118979-65-0
  • Molecular Weight : 229.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart