LIF432341900 Life Chemicals Inc.

4-(2-bromobenzyl)-1-(phenylsulfonyl)piperidine-4-carboxylic acid

  • MDL number MFCD11987064
  • Cas Number : 1171661-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart