LIF432341901 Life Chemicals Inc.

4-oxo-1,2,5,6-tetrahydro-4H-pyrrolo[3,2,1-ij]quinoline-8-sulfonyl chloride

  • MDL number MFCD19337861
  • Cas Number : 1268026-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart