LIF432341903 Life Chemicals Inc.

3-(2-fluorophenyl)pyrrolidine hydrochloride

  • MDL number MFCD03094757
  • Cas Number : 943843-62-7
  • Molecular Weight : 201.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart