LIF432341907 Life Chemicals Inc.

{[1-(6-methylpyridazin-3-yl)piperidin-4-yl]methyl}amine

  • MDL number MFCD16110375
  • Cas Number : 1250271-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart