LIF432341908 Life Chemicals Inc.

2-(3,5-dichlorophenyl)morpholine oxalate

  • MDL number MFCD03840013
  • Cas Number : 1171742-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart